missing translation for 'onlineSavingsMsg'
Learn More

CD47, Functional Grade, clone: B6H12, eBioscience™

Mouse Monoclonal Antibody

£64.00 - £232.00

Specifications

Antigen CD47
Clone B6H12
Target Species Human
Conjugate Functional Grade
Isotype IgG1, kappa, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15288457 Affymetrix eBioscience
16-0479-81
50μg £64.00
50µg
EU Stock 12
Add to basket
15298457 Affymetrix eBioscience
16-0479-85
500μg £232.00
500µg
EU Stock 9
Add to basket
Description

Description

Description: The monoclonal antibody B6H12 reacts to CD47 also known as integrin-associated protein (IAP), and neurophilin. CD47 is a glycosylated five transmembrane protein with a small alternatively spliced cytoplasmic domain. CD47 is involved in adhesion through interactions with SIRP (signal regulator protein) and is non-covalently associated with beta3 integrins CD51/CD61 and CD41/CD61. Furthermore this interaction can mediate bi-directional signaling to modify neural synaptic activity and regulate the phagocytic activities of macrophages. CD47 is the receptor for thrombospondin. T cell expression of CD47 can mediate activation or apoptosis (in the presence of high levels of thrombospondin). Recently stimulation of CD47 by monoclonal antibody has been shown to induce CD4+CD25- suppressive activity also increasing expression of Foxp3. Expression is found in the majority of hematopoietic cells including T and B cells, monocytes, platelets and erythrocytes (as part of the Rh complex). Expression is also found in non-hematopoietic cells.This antibody has been reported to have neutralizing activity.Applications Reported: This B6H12 antibody has been reported for use in flow cytometric analysis and neutralizing assays.Applications Tested: This B6H12 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Endotoxin: Less than 0.001 ng/ug antibody as determined by the LAL assay.Storage and handling: Use in a sterile environment.Filtration: 0.2μm post-manufacturing filtered.Purity: Greater than 90%, as determined by SDS-PAGE.Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay.Aggregation: Less than 10%, as determined by HPLC.
Specifications

Specifications

CD47
Human
IgG1, kappa, kappa
Monoclonal
Liquid
Antibody
4° C
B6H12
Functional Grade
Flow Cytometry, Functional Assay
Primary
Monoclonal
PBS with no preservative; pH 7.2
RUO

For Research Use Only.