missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD46 Monoclonal Antibody (MEM-258), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen A15711
Code : NEW
Additional Details : Weight : 0.00280kg
Description
CD46 (MCP, membrane cofactor protein) is a multifunctional cell surface transmembrane protein that binds and inactivates C3b and C4b complement fragments, regulates T cell-induced inflammatory responses by either inhibiting (CD46-1 isoform) or increasing (CD46-2 isoform) the contact hypersensitivity reaction. CD46 also serves as a receptor for several human pathogens (both bacteria and viruses), and its ligation alteres T lymphocyte polarization toward antigen-presenting cells or target cells, inhibiting lymphocyte function. CD46 is a protector of placental tissue and is also expressed on the inner acrosomal membrane of spermatozoa.Specifications
CD46 | |
Monoclonal | |
APC | |
CD46 | |
AHUS2, MCP, MGC26544, MIC10, TLX, TRA2.10 | |
Mouse | |
Purified | |
RUO | |
4179 | |
4° C | |
Liquid |
Flow Cytometry | |
MEM-258 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P15529 | |
CD46 | |
Human CD46 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |