missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45RO Monoclonal Antibody (UCHL1), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD45RO01
Code : 50
Additional Details : Weight : 0.02000kg
Description
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
CD45RO Monoclonal antibody specifically detects CD45RO in Human samples. It is validated for Flow Cytometry.Specifications
CD45RO | |
Monoclonal | |
FITC | |
PTPRC | |
B220; Cd45; CD45 antigen; CD45R; GP180; Lca; L-CA; leucocyte common antigen; leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; loc; LY5; Ly-5; lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; membrane tyrosine phosphatase; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Protein tyrosine phosphatase, receptor-type, c polypeptide; PTPRC; Receptor-type tyrosine-protein phosphatase C; RT7; T200; T200 glycoprotein; T200 leukocyte common antigen | |
Mouse | |
purified | |
ASR, ASR/IVD | |
5788 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
UCHL1 | |
PBS with 4mg/mL BSA and 0.1% sodium azide | |
P08575 | |
PTPRC | |
Human CD45RO | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |