missing translation for 'onlineSavingsMsg'
Learn More

CD45RO, eFluor 450, clone: UCHL1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-0457-41

Code : Z2

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15286717

  • £70.03 / 25 tests


EU Stock 8
Add to basket

Description

Description

Description: The UCHL1 monoclonal antibody reacts with human CD45RO, a 180kDa isoform of CD45. CD45RO is expressed by most thymocytes, activated memory T cells, granulocytes and monocytes. CD22 is a ligand for CD45RO. Expression of CD45RO and CD45RA is used commonly to discriminate subsets of peripheral T cells.Applications Reported: This UCHL1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This UCHL1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD45RO
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P08575
B220, CD45, CD45R, GP180, LCA, LY5, T200
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
UCHL1
eFluor 450
PTPRC
Conjugated
PTPRC
IgG2a, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
5788
Videos

For Research Use Only.