missing translation for 'onlineSavingsMsg'
Learn More

CD45RO Mouse anti-Human, APC-eFluor® 780, Clone: UCHL1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 47-0457-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15569546

  • £82.75 / 25 tests

In stock, ready to ship: 1

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 633-647 nm; Emission: 780 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. CD45RO (UCHL1, ubiquitin carboxyl terminal esterase L1) is the shortest isoform of a receptor-type protein tyrosine phosphatase, CD45 glycoprotein. CD45 is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases, promotes cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis. CD45 isoforms differ in their extracellular domains, whereas they share identical transmembrane and cytoplasmic domains. CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. CD45RO is expressed on macrophages, CD8+ T cells, activated T cells and myeloma cells. Besides the role in immunoreceptor signaling, CD45 is important in promoting cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis.
Specifications

Specifications

CD45RO
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P08575
B220, CD45, CD45R, GP180, LCA, LY5, T200
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
UCHL1
APC-eFluor™ 780
PTPRC
Liquid
PTPRC
IgG2a, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
5788
Videos
Documents

For Research Use Only.