missing translation for 'onlineSavingsMsg'
Learn More

CD45RB, Biotin, clone: C363.16A, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0455-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15209579

  • £49.53 / 100µg


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Description

Description

Description: The C363.16A monoclonal antibody reacts with the exon B-dependent isoform of mouse CD45. This CD45 isoform is expressed by thymocytes, peripheral B cells and a subpopulation of T cells at high density and by dendritic cells and macrophages at low density. CD45 is a protein tyrosine phosphatase and serves an important role in leukocyte signal transduction.Applications Reported: The C363.16A antibody has been reported for use in flow cytometric analysis.Applications Tested: The C363.16A antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD45RB
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P06800
B220, CD45, CD45R, GP180, LCA, LY5, T200
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
C363.16A
Biotin
Ptprc
Conjugated
Ptprc
IgG2a, kappa
100μg
Store at 2-8°C. Do not freeze.
Primary
19264
Videos

For Research Use Only.