missing translation for 'onlineSavingsMsg'
Learn More

CD45RA, APC, clone: OX33, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-0462-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15300630

  • £61.65 / 25µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: This OX33 monoclonal antibody reacts with rat CD45RA. In rats, this isoform of CD45, which is also known as leukocyte common antigen, is present only on B cells. CD45 is a type I transmembrane protein tyrosine phosphatase that exists as a variety of different isoforms due to alternative splicing.Applications Reported: This OX33 antibody has been reported for use in flow cytometric analysis.Applications Tested: This OX33 antibody has been tested by flow cytometric analysis of rat splenoctyes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD45RA
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P04157
B220, CD45, CD45R, GP180, LCA, LY5, T200
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Rat
Flow Cytometry
OX33
APC
Ptprc
Conjugated
Ptprc
IgG1, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
24699
Videos

For Research Use Only.