missing translation for 'onlineSavingsMsg'
Learn More

CD45RA, PE-Cyanine5, clone: HI100, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 15-0458-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15507736

  • £75.29 / 25 tests


Estimated Shipment date
from Supplier 29-06-2018
Add to basket

Description

Description

Description: The HI100 monoclonal antibody reacts with human CD45RA, a 220kDa molecule expressed by subpopulations of CD4+ peripheral T lymphocytes, CD8+ peripheral T lymphocytes, and B cells. The CD45RA+ T cell populations are mainly naive/virgin allowing the use of HI100 mAb as a phenotypic marker to discriminate T cell subsets.Applications Reported: The HI100 antibody has been reported for use in flow cytometric analysis.Applications Tested: This HI100 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-561 nm; Emission: 667 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD45RA
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P08575
B220, CD45, CD45R, GP180, LCA, LY5, T200
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
HI100
PE-Cyanine5
PTPRC
Conjugated
PTPRC
IgG2b, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
5788
Videos

For Research Use Only.