missing translation for 'onlineSavingsMsg'
Learn More

CD45RA, PE, clone: JS-83, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-9979-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15557296

  • £61.93 / 25 tests


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Description

Description

Description: The JS-83 monoclonal antibody reacts with human CD45RA, a 220kDa molecule expressed by subpopulations of CD4^+ peripheral T lymphocytes, CD8^+ peripheral T lymphocytes, and B cells. The CD45RA^+ T cell populations are mainly naive/virgin allowing the use of JS-83 mAb as a phenotypic marker to discriminate T cell subsets.Applications Reported: JS-83 has been reported for use in flow cytometric analysis.Applications Tested: This JS-83 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.03 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD45RA
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P08575
B220, CD45, CD45R, GP180, LCA, LY5, T200
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
JS-83
PE
PTPRC
Conjugated
PTPRC
IgG1, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
5788
Videos

For Research Use Only.