Learn More
Epredia™ Lab Vision™ CD45RA Ab-3 Mouse Monoclonal Antibody, 1mL; 200μg/mL, BSA and azide
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD45RA Ab-3, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-1845-P
Code : LD
Additional Details : Weight : 0.00100kg
Description
CD45 or Leukocyte Common Antigen (LCA/T200) belongs to a family of membrane sialoglycoproteins with MW of 180kDa-220 kDa. Human CD45 contains three exons which encode peptide segments designated A, B and C, respectively. The differential splicing of the exons generates various isoforms. CD45RA.is a single chain glycoprotein. It is expressed on appoximately 40-50% of peripheral CD4+ T-cells, 50% of peripheral CD8+ T-cells, B-cells, and leukemic B-cell lines. T-cells expressing CD45RA and CD45RO define complementary, predominantly non-overlapping populations of resting peripheral T-cells. CD45RA are naive or virgin T-cells while, T-cells expressing CD45RO are memory T-cells.Host Species: Mouse
Clone: 4KB5
Isotype: IgG1/κ
Species Reactivity: Human. Others not tested
Epitope: Not determined
Immunogen: Hairy cell leukemia cells
Molecular Weight: 205-220 kDa
Positive Control: Lymph node or tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
CD45RA Ab-3 | |
Monoclonal | |
200μg/mL | |
Mouse | |
1 mL | |
Oncogenesis | |
Human | |
IgG2a κ |
Immunohistochemistry (Paraffin) | |
4KB5 | |
Unconjugated | |
Hairy cell leukemia cells | |
IVD | |
Primary | |
Concentrated |