missing translation for 'onlineSavingsMsg'
Learn More

CD45R (B220) Rat anti-Human, Mouse, PerCP-eFluor 710, Clone: RA3-6B2, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-0452-82

UNSPSC : 12352200

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15331260

  • £189.20 / 100µg


EU Stock 1
Add to basket

Disclaimers:

For Research Use Only.


Specifications

Description

The RA3-6B2 monoclonal antibody reacts with exon A-restricted isoform of mouse CD45, a 220kDa surface molecule. CD45R/B220 epitope is mainly expressed by the B cell lineage from early Pro-B to mature B cells. However, some activated T cells, lymphokine activated killer cells (LAK), NK cell progenitors in the bone marrow, and T cells of the \I{lpr/lpr\I} mutant mouse also express this antigen.
Description & Specifications

Specifications

CD45R (B220)
RA3-6B2
PerCP-eFluor 710
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Rat
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
Conjugated
Ly-5, Lyt-4, T200
IgG2a
RUO
Primary
Murine