missing translation for 'onlineSavingsMsg'
Learn More

CD45R (B220) Rat anti-Human, Mouse, eFluor 660, Clone: RA3-6B2, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-0452-80

UNSPSC : 12352200

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15331870

  • £54.42 / 25µg


Estimated Shipment date
from Supplier 22-09-2017
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

The RA3-6B2 monoclonal antibody reacts with exon A-restricted isoform of mouse CD45, a 220kDa surface molecule. CD45R/B220 epitope is mainly expressed by the B cell lineage from early Pro-B to mature B cells. However, some activated T cells, lymphokine activated killer cells (LAK), NK cell progenitors in the bone marrow, and T cells of the \I{lpr/lpr\I} mutant mouse also express this antigen.
Specifications

Specifications

CD45R (B220)
RA3-6B2
eFluor 660
Conjugated
Rat
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Immunohistochemistry (Frozen)
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Ly-5, Lyt-4, T200
IgG2a
RUO
Primary
Murine