missing translation for 'onlineSavingsMsg'
Learn More

CD45R (B220), PE, clone: YKIX753.22, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-5451-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15557006

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

The YKIX753.22 monoclonal antibody reacts with canine CD45R. In dogs, CD45R is expressed on B cells and peripheral T cells. In addition, CD45 exists as multiple isoforms due to alternative splicing. Some isoforms of CD45 are CD45RA, CD45RB, or CD45RO depending on the exon used during transcription. The C-terminal domain of CD45 exhibits tyrosine phosphatase activity that enables signaling from by T cell and B cell receptors following activation.

This antibody has been reported to promote cell depletion via antibody-dependent cell-mediated cytotoxicity when used in conjunction with YKIX716.13.
Specifications

Specifications

CD45R (B220)
YKIX753.22
PE
Conjugated
IgG2b, kappa
RUO
Primary
Canine
Flow Cytometry
5μL (0.0075μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Rat
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Videos

For Research Use Only.