missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45, FITC, clone: 30-F11, eBioscience™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 11-0451-85
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The 30-F11 monoclonal antibody reacts with all isoforms of mouse CD45, also known as Leukocyte Common Antigen (LCA). CD45 is expressed by all hematopoietic cells excluding mature erythrocytes and platelets. The cytoplasmic portion of CD45 has tyrosine phosphatase enzymatic activity and plays an important role in activation of lymphocytes.Applications Reported: The 30-F11 antibody has been reported for use in flow cytometric analysis.Applications Tested: The 30-F11 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.Specifications
CD45 | |
Monoclonal | |
0.5mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
P06800 | |
Cluster designation 45, common leukocyte antigen, T220 and B220 | |
Rat | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Murine |
Flow Cytometry | |
30-F11 | |
FITC | |
Ptprc | |
Conjugated | |
Ptprc | |
IgG2b, kappa | |
500μg | |
Store at 2-8°C. Do not freeze. Light-sensitive material. | |
Primary | |
19264 |
For Research Use Only.