missing translation for 'onlineSavingsMsg'
Learn More

CD45, Biotin, clone: 30-F11, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0451-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15269559

  • £30.77 / 50µg


EU Stock 13
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The 30-F11 monoclonal antibody reacts with all isoforms of mouse CD45, also known as Leukocyte Common Antigen (LCA). CD45 is expressed by all hematopoietic cells excluding mature erythrocytes and platelets. The cytoplasmic portion of CD45 has tyrosine phosphatase enzymatic activity and plays an important role in activation of lymphocytes.Applications Reported: The 30-F11 antibody has been reported for use in flow cytometric analysis.Applications Tested: The 30-F11 antibody has been tested by flow cytometric analysis of mouse bone marrow cells and splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD45
30-F11
Biotin
Conjugated
Rat
50μg
Store at 2-8°C. Do not freeze.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Leukocyte Common Antigen, LCA, Ly-5
IgG2b
RUO
Primary
Murine
Videos