missing translation for 'onlineSavingsMsg'
Learn More

CD45, APC-eFluor 780, clone: 30-F11, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 47-0451-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15391620

  • £110.65 / 100µg


EU Stock 26
Add to basket

Description

Description

Description: The 30-F11 monoclonal antibody reacts with all isoforms of mouse CD45, also known as Leukocyte Common Antigen (LCA). CD45 is expressed by all hematopoietic cells excluding mature erythrocytes and platelets. The cytoplasmic portion of CD45 has tyrosine phosphatase enzymatic activity and plays an important role in activation of lymphocytes.Applications Reported: This 30-F11 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 30-F11 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.APC-eFluor® emits at 780 nm and is excited with the Red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Light sensitivity: This tandem is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 633-647 nm; Emission: 780 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD45
Monoclonal
0.2mg/mL
PBS with 0.09% sodium azide; pH 7.2
P06800
Cluster designation 45, common leukocyte antigen, T220 and B220
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
30-F11
APC-eFluor 780
Ptprc
Conjugated
Ptprc
IgG2b, kappa
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Primary
19264
Videos

For Research Use Only.