missing translation for 'onlineSavingsMsg'
Learn More

CD45 Rat anti-Canine, eFluor® 450, Clone: YKIX716.13, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-5450-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15599756

  • £65.50 / 25 tests

Estimated Shipment
03-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

2 μm post- manufacturing filtered. CD45 (LCA, leukocyte common antigen) is a receptor-type protein tyrosine phosphatase (PTP) ubiquitously expressed in all nucleated hematopoietic cells, comprising approximately 10% of all surface proteins in lymphocytes. CD45 is absent on non-hematopoietic cell lines, normal and maligt, non-hematopoietic tissues. CD45 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45 protein exists as multiple isoforms as a result of alternative splicing, differ in their extracellular domains but share identical transmembrane and cytoplasmic domains. CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells. CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. CD45 has been shown to be an essential regulator of T- and B-cell antigen receptor signaling and suppresses JAK kinases to regulate cytokine receptor signaling. CD45 is also important in promoting cell survival by modulating integrin-mediated signal transduction pathway, DNA fragmentation during apoptosis and inhibition or upregulation of various immunological functions.
Specifications

Specifications

CD45
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Liquid
PTPRC
IgG2b, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
490255
Flow Cytometry
YKIX716.13
eFluor™ 450
PTPRC
Cluster designation 45; common leukocyte antigen; T220 and B220; HCD45
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Canine
Videos
Documents

For Research Use Only.