missing translation for 'onlineSavingsMsg'
Learn More

CD45 Mouse, Qfluor 710, Clone: HI30, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB46-QF710-25Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15962088

  • £59.47 / 25 tests
Estimated Shipment: 17-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD45
HI30
Qfluor 710
NP_002829
Mouse
Protein A/G Purified
RUO
Primary
5788
Flow Cytometry
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
PTPRC
IgG1 kappa
25 Tests
Store at 2-4°C.
Monoclonal
Videos
SDS
Documents
Certificates
Special Offers

Special Offers