missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Mouse, Qfluor 710, Clone: HI30, enQuireBio™
Mouse Monoclonal Antibody
£62.45 - £109.00
Specifications
Antigen | CD45 |
---|---|
Clone | HI30 |
Concentration | 5μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15962088
|
EnquireBio
QAB46-QF710-25TESTS |
25 Tests |
£62.45
25 tests |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15952088
|
EnquireBio
QAB46-QF710-100TESTS |
100 Tests |
£109.00
100 tests |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD45 | |
5μL/Test | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
5788 | |
IgG1 κ | |
Protein A/G Purified |
HI30 | |
Flow Cytometry | |
Qfluor 710 | |
RUO | |
NP_002829 | |
PTPRC | |
Primary | |
Store at 2-4°C. |