missing translation for 'onlineSavingsMsg'
Learn More

CD45, PE, clone: OX1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0461-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15536786

  • £124.73 / 100µg


Estimated Shipment date
from Supplier 03-04-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This OX1 monoclonal antibody recognizes a non-polymorphic determinant of rat CD45, which is also known as the leukocyte common antigen. CD45 is expressed on all hematopoietic cells except erythrocytes and platelets. This type I transmembrane protein tyrosine phosphatase exists as a variety of different isoforms due to alternative splicing.Applications Reported: This OX1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This OX1 antibody has been tested by flow cytometric analysis of rat splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD45
OX1
PE
Conjugated
Murine
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Leukocyte Common Antigen
IgG1
RUO
Primary
Rat
Videos