missing translation for 'onlineSavingsMsg'
Learn More

CD45, APC, clone: OX1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-0461-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15380620

  • £56.04 / 25µg


Estimated Shipment
04-07-2018
Add to basket

Description

Description

Description: This OX1 monoclonal antibody recognizes a non-polymorphic determinant of rat CD45, which is also known as the leukocyte common antigen. CD45 is expressed on all hematopoietic cells except erythrocytes and platelets. This type I transmembrane protein tyrosine phosphatase exists as a variety of different isoforms due to alternative splicing.Applications Reported: This OX1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This OX1 antibody has been tested by flow cytometric analysis of rat splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD45
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P04157
Cluster designation 45, common leukocyte antigen, T220 and B220
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Rat
Flow Cytometry
OX1
APC
Ptprc
Conjugated
Ptprc
IgG1, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
24699
Videos

For Research Use Only.