Learn More
CD45 Monoclonal Antibody (HI30), TRI-COLOR™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD45065
717.36 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 50
Additional Details : Weight : 0.02000kg
Description
The TRI-COLOR® conjugate is a tandem R-phycoerythrin (PE)-Cy5® conjugate that permits simultaneous multicolor labeling and detection of multiple targets with excitation by a single excitation source-the 488 nm line of the argon-ion laser.
CD45 Monoclonal antibody specifically detects CD45 in Human samples. It is validated for Flow Cytometry.Specifications
CD45 | |
Monoclonal | |
TRI-COLOR | |
PTPRC | |
B220; cd45; CD45 antigen; CD45 antigen isoform 1 precursor; CD45 antigen isoform 2 precursor; CD45 antigen isoform 3 precursor; CD45 antigen isoform 4 precursor; CD45 antigen isoform 5 precursor; CD45 antigen isoform 6 precursor; CD45R; GP180; Lca; L-CA; leucocyte common antigen; leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; leukocyte common antigen, CD45; loc; LOW QUALITY PROTEIN: receptor-type tyrosine-protein phosphatase C; LY5; Ly-5; lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; membrane tyrosine phosphatase; protein tyrosine phosphatase lambda; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Protein tyrosine phosphatase, receptor-type, c polypeptide; protein tyrosine phosphatase; alternatively spliced; Ptprc; Receptor-type tyrosine-protein phosphatase C; RT7; T200; T200 glycoprotein; T | |
Mouse | |
purified | |
RUO, ASR/IVD | |
5788 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
HI30 | |
PBS with BSA, sucrose and 0.1% sodium azide | |
P08575 | |
PTPRC | |
CD45 antigen | |
2.5 mL | |
Primary | |
Human | |
Antibody | |
IgG1 |