missing translation for 'onlineSavingsMsg'
Learn More

CD45, PE, clone: HI30, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0459-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15506786

  • £54.42 / 25 tests


EU Stock 3
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The HI30 monoclonal antibody reacts with all isoforms of human CD45, also known as Leukocyte Common Antigen (LCA). CD45 is expressed by all hematopoietic cells excluding circulating erythrocytes and platelets. The cytoplasmic portion of CD45 has tyrosine phosphatase enzymatic activity and plays an important role in activation of lymphocytes.Applications Reported: The HI30 antibody has been reported for use in flow cytometric analysis.Applications Tested: This HI30 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD45
HI30
PE
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.06μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Leukocyte Common Antigen, LCA, Ly-5
IgG1
RUO
Primary
Human
Videos