missing translation for 'onlineSavingsMsg'
Learn More

CD45, FITC, clone: 2D1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-9459-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15506626

  • £146.84 / 100 tests


EU Stock 25
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The 2D1 monoclonal antibody reacts with all isoforms of human CD45, also known as Leukocyte Common Antigen (LCA). CD45 is expressed by all hematopoietic cells excluding circulating erythrocytes and platelets. The cytoplasmic portion of CD45 has tyrosine phosphatase enzymatic activity and plays an important role in activation of lymphocytes.Applications Reported: This 2D1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 2D1 antibody has been pre-titrated and tested by flow cytometric analysis of human peripheral blood leukocytes. This can be used at 5 μL (1.0 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD45
2D1
FITC
Conjugated
Murine
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (1.0μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
LCA
IgG1
RUO
Primary
Human
Videos