missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45.2 Mouse, Qfluor 710, Clone: 104, enQuireBio™
Mouse Monoclonal Antibody
£89.95 - £191.00
Specifications
Antigen | CD45.2 |
---|---|
Clone | 104 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15952038
|
EnquireBio
QAB43-QF710-25UG |
25 μg |
£89.95
25µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15942038
|
EnquireBio
QAB43-QF710-100UG |
100 μg |
£191.00
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD45.2 | |
0.5mg/mL | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
19264 | |
IgG2a κ | |
Protein A/G Purified |
104 | |
Flow Cytometry | |
Qfluor 710 | |
RUO | |
NP_001255215 | |
Ptprc | |
Primary | |
Store at 2-4°C. |