CD45.2 Mouse anti-Mouse, Clone: 104, eBioscience™

Mouse Monoclonal Antibody

Overview
Brand: Affymetrix eBioscience

Manufacturer Part Number: 14-0454-81

UNSPSC: 12352200

Code: Z2

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only.

Product Code. 15206977

Quantity Price
1 £ 36.08 / 50µg
Estimated Shipment date
from Supplier 31-05-2017
Add to basket


Description and Specification

Specification

Antigen CD45.2
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Clone 104
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Purified
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Gene Alias C57BL/6, Balb/c, Ly5.2
Host Species Murine
Isotype IgG2a
Quantity 50μg
Regulatory Status RUO
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Target Species Murine

The 104 monoclonal antibody reacts with the mouse CD45 molecule, the leukocyte common antigen (LCA) in CD45.2-expressing mouse strains. The strains that express CD45.2 include the most commonly used mouse strains C57BL/6, BALB/c, C58, DBA/1, DBA/2, C3H/He, CBA, 129, A and AKR. CD45.2 is expressed by all leukocytes in these strains.