missing translation for 'onlineSavingsMsg'
Learn More

CD45.1 Mouse, PE-Cy7, Clone: A20, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB42-PE7-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15942018

  • £168.00 / 100µg

Estimated Shipment
30-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD45.1
A20
PE-Cyanine7
NP_001255215
Mouse
Protein A/G Purified
RUO
Primary
19264
Flow Cytometry
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Ptprc
IgG2a kappa
100 μg
Store at 2-4°C.
Monoclonal
Videos
SDS
Documents