missing translation for 'onlineSavingsMsg'
Learn More

CD45.1 Mouse anti-Mouse, PE-eFluor® 610, Clone: A20, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 61-0453-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15580437

  • £79.00 / 25µg

Estimated Shipment
07-12-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. CD45 (LCA, leukocyte common antigen) is a receptor-type protein tyrosine phosphatase ubiquitously expressed in all nucleated hematopoietic cells, comprising approximately 10% of all surface proteins in lymphocytes. CD45 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45 protein exists as multiple isoforms as a result of alternative splicing; these isoforms differ in their extracellular domains, whereas they share identical transmembrane and cytoplasmic domains. These CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. Besides the role in immunoreceptor signaling, CD45 is important in promoting cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis. CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells.
Specifications

Specifications

CD45.1
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P06800
CD45, L-CA, Lca, RT7, T200
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
A20
PE-eFluor™ 610
Ptprc
Liquid
Ptprc
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
19264
Videos
SDS
Documents

For Research Use Only.