missing translation for 'onlineSavingsMsg'
Learn More

CD45.1, Biotin, clone: A20, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0453-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15239569

  • £49.65 / 50µg


EU Stock 2
Add to basket

Description

Description

Description: The A20 monoclonal antibody reacts with the mouse CD45 molecule, the leukocyte common antigen (LCA) in CD45.1-expressing mouse strains. The strains that express CD45.1 include SJL/J, DA, STS/A and RIII. CD45.1 is expressed by all leukocytes in these strains.Applications Reported: The A20 antibody has been reported for use in flow cytometric analysis, and immunohistochemical staining of frozen tissue sections.Applications Tested: The A20 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD45.1
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P06800
CD45, L-CA, Lca, RT7, T200
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
A20
Biotin
Ptprc
Conjugated
Ptprc
IgG2a, kappa
50μg
Store at 2-8°C. Do not freeze.
Primary
19264
Videos

For Research Use Only.