missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD44 Std./H-CAM Ab-4 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunoprecipitation and flow cytometry experiments.
£98.23 - £442.00
Specifications
Antigen | CD44 Std./H-CAM Ab-4 |
---|---|
Clone | 156-3C11 |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Host Species | Mouse |
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
12661977
|
Epredia
MS-668-B1 |
Concentrated | Biotin | 500 μL |
£433.00
500µL |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12601987
|
Epredia
MS-668-P1ABX |
Concentrated | Unconjugated | 100 μL |
£329.00
100µL |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12691977
|
Epredia
MS-668-P1 |
Concentrated | Unconjugated | 500 μL |
£294.00
500µL |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12671977
|
Epredia
MS-668-P |
Concentrated | Unconjugated | 1 mL |
£442.00
1mL |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12611987
|
Epredia
MS-668-PABX |
Concentrated | Unconjugated | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12681977
|
Epredia
MS-668-P0 |
Concentrated | Unconjugated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12651977
|
Epredia
MS-668-B0 |
Concentrated | Biotin | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12651987
|
Epredia
MS-668-R7 |
Prediluted | Unconjugated | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
CD44 is a cell surface glycoprotein expressed on lymphocytes, monocytes, and granulocytes. CD44 is also known as homing cell adhesion molecule (H-CAM), Phagocytic glycoprotein-1 (PgP-1), ECM-III, HUTCH-1, or Hermes-1. CD44 has been implicated in cell migration, in lymphocyte homing, and in tumor metastasis. Ab-4 reacts with red cells pretreated with 2-aminoethylisothiouronium bromide (AET). Digestion of intact erythrocytes by trypsin treatment leaves a 50-54 kDa protein fragment attached to the membrane, while chymotrypsin leaves a somewhat similarly sized 51-56 kDa component that are recognized by Ab-4. This antibody enhances IL-2 production by CD2-transfected murine hybridoma stimulated by JY cells while it has no significant effect on IL-2 production by Jurkat cells stimulated with CD3, PMA, or CD2. Ab-4 enhances proliferation of PBL stimulated with CD2.Host Species: Mouse
Clone: 156-3C11
Isotype: IgG2a
Species Reactivity: Human, Baboon and Green Monkey. Does not react with rat. Others not tested.
Epitope: Resistant to digestion by trypsin and chymotrypsin
Immunogen: Stimulated human leukocytes
Molecular Weight: 80-95 kDa
Positive Control: HUVEC or HeLa cells, tonsil or esophageal CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Native verified)
- Flow Cytometry
Specifications
CD44 Std./H-CAM Ab-4 | |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Mouse | |
Oncogenesis | |
Stimulated human leukocytes | |
Primary |
156-3C11 | |
Monoclonal | |
IVD | |
Human, Non-human Primate | |
IgG2a |