missing translation for 'onlineSavingsMsg'
Learn More

CD44, PE, clone: IM7, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0441-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15208539

  • £48.31 / 50µg


EU Stock 5
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The IM7 monoclonal antibody reacts with all isoforms of mouse CD44 (Pgp-1). CD44 is expressed by hematopoietic and non-hematopoietic cells. Bone marrow myeloid cells and memory T cells highly express this antigen and peripheral B and T cells can upregulate the expression of CD44. CD44 functions as an adhesion molecule through its binding to hyaluronate, an extracellular matrix component.Applications Reported: The IM7 antibody has been reported for use in flow cytometric analysis.Applications Tested: The IM7 antibody has been tested by flow cytometric analysis of mouse bone marrow cells and splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD44
IM7
PE
Conjugated
Rat
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Pgp-1, MDU3, Hermes, Hyaluronate receptor
IgG2b
RUO
Primary
Murine
Videos