Learn More
CD44 Monoclonal Antibody (YKIX337.8), FITC, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£67.25 - £206.00
Specifications
Antigen | CD44 |
---|---|
Clone | YKIX337.8 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15576556
|
Affymetrix eBioscience
11-5440-42 |
100 Tests |
£206.00
100 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15566556
|
Affymetrix eBioscience
11-5440-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: This YKIX337.8 monoclonal antibody reacts with canine CD44, also known as phagocytic glycoprotein-1 (Pgp-1). CD44 is an adhesion molecule that binds hyaluronan, a major component of the extracellular matrix. This cell surface molecule is expressed on leukocytes and epithelial cells, as well as in canine mammary and melanocytic tumors. Furthermore, CD44 expression is induced by T cell activation. Applications Reported: This YKIX337.8 antibody has been reported for use in flow cytometric analysis. Applications Tested: This YKIX337.8 antibody has been pre-titrated and tested by flow cytometric analysis of normal canine peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD44 cell surface antigen is a 100 kDa type 1 transmembrane glycoprotein widely expressed on human leucocytes, white matter of the brain and by some epithelial cells of the intestine and breast. Several isoforms of CD44 exist, including the predominant CD44H isoform detected in many normal tissues. CD44 is a receptor for hyaluronic acid (HA) and is involved in cell-cell interactions, cell adhesion and migration. CD44 also participates in a wide variety of cellular functions including lymphocyte activation, recirculation and homing. CD44 expression may be up-regulated upon some carcinomas, and it has been speculated that this may be related to metastatic potential. CD44 is expressed by hematopoietic, non-hematopoietic cells, epithelial tissues, and to filopodia in cultured keratinocytes. Further, bone marrow myeloid cells and memory T cells express CD44 at high levels, and peripheral B and T cells can upregulate the expression of CD44 in response to certain stimulatory events. Transcripts for the CD44 gene undergo complex alternative splicing that results in many functionally distinct isoforms, however, the full-length nature of some of these variants have not been determined. Alternative splicing is the basis for the structural and functional diversity of the CD44 protein. Diseases associated with CD44 dysfunction include superficial keratitis and lichen sclerosus. CD44 also may be related to tumor metastasis formation.Specifications
CD44 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
Q28284 | |
CD44 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CD44 |
YKIX337.8 | |
Flow Cytometry | |
FITC | |
Rat | |
Canine | |
403939 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.