missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CD44 Pre-design Chimera RNAi

Homo sapiens CD44 antigen (homing function and Indian blood group system) (CD44), transcript variant 8, mRNA.

Brand:  Abnova™ H00000960-R12.10nmol

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16003828

  • £172.47 / 10nmol

Estimated Shipment
02-10-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Gene name: CD44
  • Gene Alias: CDW44,CSPG8,ECMR-III,HCELL,IN,LHR,MC56,MDU2,MDU3,MGC10468,MIC4,MUTCH-I,Pgp1
  • Entrez GeneID: 960
  • strong>Gene Description: CD44 molecule (
Specifications

Specifications

RNAi Knockdown
DEPC water
10 nmol
Human
Homo sapiens CD44 antigen (homing function and Indian blood group system) (CD44), transcript variants, mRNA.
CD44 Pre-design Chimera RNAi
Store at -20°C, do not exceed 4 - 5 freeze-thaw cycles to ensure product integrity.
Videos
Documents

For research use only