missing translation for 'onlineSavingsMsg'
Learn More

CD43 Monoclonal Antibody (eBioR2/60), FITC, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-0431-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15278079

  • £124.00 / 100µg
Estimated Shipment: 29-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The eBioR2/60 monoclonal antibody reacts with mouse CD43, also known as leukosialin and sialophorin. CD43 has glycoforms of approximately 115 and 130 kDa, and is expressed by an early B cell subset in BM, thymocytes and mature T cells, NK cells, myeloid lineage cells and platelets. CD43 is reported to bind to CD54 and play a role in adhesion and B cell survival. While the S7 and 1B11 monoclonal antibody clones detect only certain isoforms of CD43, data suggests that R2/60 detects the expression of both isoforms. It has been demonstrated that R2/60 stains the majority of CD4+ and CD8+ splenocytes, as well as a significant population of CD19+ splenocytes. However, the clones S7 and 1B11 stain only CD4+ and CD8+, or CD19+ splenocytes, respectively. This suggests that S7 and 1B11 recognize distinct epitopes which are differentially expressed by splenic B and T cells, whereas R2/60 recognizes an epitope shared by both isoforms of CD43. Applications Reported: This eBioR2/60 antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioR2/60 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

CD43 (leukosialin, sialophorin) is a transmembrane mucin-like protein with high negative charge, expressed on the surface of most hematopoietic cells. CD43 contributes to a repulsive barrier that interferes with cellular adhesion, however, in certain cases also promotes leukocyte aggregation. By interaction with actin-binding proteins ezrin and moesin CD43 plays a regulatory role in remodeling T-cell morphology and regulates cell-cell interactions during lymphocyte traffic. CD43 signaling both enhances LFA-1 adhesiveness and counteracts LFA-1 induction via other receptors. Expression of CD43 causes induction of functionally active tumor suppressor p53 protein, but in case of p53 and ARF deficiency CD43 promotes tumor proliferation and viability. CD43 appears to be an important modulator of leukocyte functions. Diseases associated with CD43 dysfunction include Wiscott-Aldrich Syndrome and Adenoid Basal Cell Carcinoma.
Specifications

Specifications

CD43
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P15702
SPN
Affinity chromatography
RUO
20737
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
eBioR2/60
FITC
SPN
3E8 antigen; A630014B01Rik; B-cell differentiation antigen LP-3; CD43; CD43 cytoplasmic tail; CD43-ct; Galactoglycoprotein; GALGP; gpL115; Leukocyte sialoglycoprotein; leukosialin; leukosianin; LOC101120462; LOW QUALITY PROTEIN: leukosialin; Lsn; Lsn1; Ly48; Ly-48; Lymphocyte antigen 48; sialophorin; sialophorin (gpL115, leukosialin, CD43); sialophorin gpL115; SPN; W3/13 antigen
Rat
100 μg
Primary
Mouse
Antibody
IgM
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.