missing translation for 'onlineSavingsMsg'
Learn More

CD42d, APC, clone: 1C2, eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-0421-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15340610

  • £193.08 / 100µg


EU Stock 5
Add to basket

Description

Description

Description: The monoclonal antibody 1C2 recognizes mouse and rat CD42d, a single transmembrane protein also known as platelet glycoprotein V (GPV). CD42d interacts with CD42a, CD42b and CD42c to form the receptor for von willebrand factor and thrombin. This functional complex plays a role in platelet adhesion. Expression of CD42d (unlike other markers such as CD41) is restricted to resting platelets and megakaryocytes and therefore is a good marker for development of megakaryocytes.Applications Reported: This 1C2 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 1C2 antibody has been tested by flow cytometric analysis of mouse platelets. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD42d
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
O08770, O08742
Platelets GP V, CD42d, Glycoprotein 5, GPV
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Rat
Flow Cytometry
1C2
APC
Gp5
Conjugated
Gp5
IgG
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
14729
Videos

For Research Use Only.