missing translation for 'onlineSavingsMsg'
Learn More

CD42b, FITC, clone: HIP1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-0429-41

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15576396

  • £59.25 / 25 tests

Estimated Shipment
30-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The HIP1 monoclonal antibody reacts with the human CD42b molecule, a 135kDa transmembrane protein also known as platelet gpIb.Applications Reported: The HIP1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This HIP1 antibody has been pre-titrated and tested by flow cytometric analysis of human platelets. This can be used at 5 μL (1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD42b
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P07359
GPIb alpha, GP1Ba
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
HIP1
FITC
GP1BA
Liquid
GP1BA
IgG1, kappa, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
2811
Videos
Documents

For Research Use Only.