missing translation for 'onlineSavingsMsg'
Learn More

CD42b, clone: HIP1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-0429-82

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15296947

  • £68.45 / 100µg


Estimated Shipment
28-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The HIP1 monoclonal antibody reacts with the human CD42b molecule, a 135kDa transmembrane protein also known as platelet gpIb.Applications Reported: The HIP1 antibody has been reported for use in flow cytometric analysis, in vitro blocking of human CD42b, immunoblotting (WB), and immunohistochemical staining. (Please use Functional Grade purified HIP1, cat. 16-0429, in functional assays.).Applications Tested: The HIP1 antibody has been tested by flow cytometric analysis of human platelets and can be used less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibodies be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD42b
HIP1
Unconjugated
Purified
IgG1
RUO
Primary
Human
Western Blot
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
100μg
Store at 2-8°C.
Monoclonal
Videos