missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD41a Monoclonal Antibody (HIP8), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£98.25 - £226.00
Specifications
Antigen | CD41a |
---|---|
Clone | HIP8 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PerCP-eFluor 710 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15509126
|
Affymetrix eBioscience
46-0419-42 |
100 Tests |
£226.00
100 tests |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15599116
|
Affymetrix eBioscience
46-0419-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
ITGA2B encodes integrin alpha chain 2b. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. Alpha chain 2b undergoes post-translational cleavage to yield disulfide-linked light and heavy chains that join with beta 3 to form a fibronectin receptor expressed in platelets that plays a crucial role in coagulation. Mutations that interfere with this role result in thrombasthenia. In addition to adhesion, integrins are known to participate in cell-surface mediated signalling.
CD41a Monoclonal antibody specifically detects CD41a in Human samples. It is validated for Flow Cytometry.Specifications
CD41a | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Mouse | |
Human | |
3674 | |
Primary | |
Antibody |
HIP8 | |
Monoclonal | |
Liquid | |
RUO | |
P08514 | |
IgG1 κ | |
Affinity chromatography |
For Research Use Only.