missing translation for 'onlineSavingsMsg'
Learn More

CD41a, PE, clone: HIP8, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0419-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15536776

  • £144.39 / 100 tests


EU Stock 1
Add to basket

Description

Description

The HIP8 monoclonal antibody reacts with the human CD41 molecule, the integrin alpha IIb also known as platelet GPIIb. CD41 non-covalently associates with integrin beta3 (GPIIIa, CD61) and is expressed by megakaryocytes and platelets. The CD41/CD61 complex is a receptor for fibronectin, fibrinogen, von Willebrand factor, vitronectin and thrombospondin and mediates platelets aggregation. HIP8 blocks platelet aggregation.
Specifications

Specifications

CD41a
HIP8
PE
Conjugated
Murine
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.125μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Integrin alpha Iib, platelet GPIIb
IgG1, kappa
RUO
Primary
Human
Videos

For Research Use Only.