missing translation for 'onlineSavingsMsg'
Learn More

CD41a, eFluor 450, clone: HIP8, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-0419-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15549676

  • £195.95 / 100 tests


EU Stock 3
Add to basket

Description

Description

Description: The HIP8 monoclonal antibody reacts with the human CD41 molecule, the integrin alpha IIb also known as platelet GPIIb. CD41 non-covalently associates with integrin beta3 (GPIIIa, CD61) and is expressed by megakaryocytes and platelets. The CD41/CD61 complex is a receptor for fibronectin, fibrinogen, von Willebrand factor, vitronectin and thrombospondin and mediates platelets aggregation. HIP8 blocks platelet aggregation.Applications Reported: This HIP8 antibody has been reported for use in flow cytometric analysis.Applications Tested: This HIP8 antibody has been pre-titrated and tested by flow cytometric analysis of human platelets. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD41a
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P08514
BDPLT2, CD41, CD41B, GP2B, GPIIb, GT, GTA, HPA3, GPalpha IIb, integrin alpha-IIb, platelet fibrinogen receptor, alpha subunit, platelet membrane glycoprotein IIb, platelet-specific antigen BAK, ITGA2B
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
HIP8
eFluor 450
ITGA2B
Conjugated
ITGA2B
IgG1, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
3674
Videos

For Research Use Only.