Learn More
CD40 Monoclonal Antibody (1C10), Super Bright™ 436, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£129.00 - £305.00
Specifications
Antigen | CD40 |
---|---|
Clone | 1C10 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15963374
|
Affymetrix eBioscience
62-0401-82 |
100 μg |
£305.00
100µg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15953374
|
Affymetrix eBioscience
62-0401-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The 1C10 monoclonal antibody reacts with mouse CD40, a 45-50 kDa type I transmembrane glycoprotein. CD40 is a member of the TNFR family and is expressed by mouse B lymphocytes, follicular dendritic cells, thymic epithelium, and a subset of peripheral T cells. CD40 regulates B cell development/maturation by inducing Ig isotype switching and in combination with other signals such as IL-4, protects B cells from surface Ig-induced apoptosis and promotes proliferation. Interaction of CD40 with CD154 (gp39), its ligand on T cells, is important in T-B cell crosstalk and plays a role in costimulation and immune regulation. The monoclonal antibody 1C10 is reported to have agonistic activity in vitro and in vivo. Applications Reported: The 1C10 antibody has been reported for use in flow cytometric analysis. Applications Tested: The 1C10 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent.
CD40 Monoclonal antibody specifically detects CD40 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD40 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
P27512 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
1C10 | |
Flow Cytometry | |
Super Bright 436 | |
Rat | |
Mouse | |
21939 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |