missing translation for 'onlineSavingsMsg'
Learn More

CD40, PE, clone: 1C10, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0401-85

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15218529

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

The 1C10 monoclonal antibody reacts with mouse CD40, a 45-50kDa type I transmembrane glycoprotein. CD40 is a member of the TNFR family and is expressed by mouse B lymphocytes, follicular dendritic cells, thymic epithelium, and a subset of peripheral T cells. CD40 regulates B cell development/maturation by inducing Ig isotype switching and in combination with other signals such as IL-4, protects B cells from surface Ig-induced apoptosis and promotes proliferation. Interaction of CD40 with CD154 (gp39), its ligand on T cells, is important in T-B cell crosstalk and plays a role in costimulation and immune regulation.

The monoclonal antibody 1C10 is reported to be agonistic in vitro and in vivo.
Specifications

Specifications

CD40
1C10
PE
Conjugated
Rat
500μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
TNFRSF5
IgG2a, kappa
RUO
Primary
Murine
Videos

For Research Use Only.