Learn More
CD40 Rat anti-Mouse, Clone: 1C10, eBioscience™
Rat Monoclonal Antibody
Brand:  Affymetrix eBioscience 14-0401-82
UNSPSC : 12352200
Code : Z2
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15226937
  • £78.88 / 100µg
Estimated Shipment
30-10-2017
Add to basket
Disclaimers:
For Research Use Only.
Description
Description
The 1C10 monoclonal antibody reacts with mouse CD40, a 45-50kDa type I transmembrane glycoprotein. CD40 is a member of the TNFR family and is expressed by mouse B lymphocytes, follicular dendritic cells, thymic epithelium, and a subset of peripheral T cells. CD40 regulates B cell development/maturation by inducing Ig isotype switching and in combination with other signals such as IL-4, protects B cells from surface Ig-induced apoptosis and promotes proliferation. Interaction of CD40 with CD154 (gp39), its ligand on T cells, is important in T-B cell crosstalk and plays a role in costimulation and immune regulation.
The monoclonal antibody 1C10 is reported to have agonistic activity in vitro and in vivo.
Specifications
Specifications
CD40
1C10
Unlabeled
Purified
Rat
100μg
Store at 2-8°C.
Monoclonal
Immunoprecipitation
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
TNFRSF5
IgG2a
RUO
Primary
Murine