missing translation for 'onlineSavingsMsg'
Learn More

CD40, PE, clone: 5C3, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0409-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15516776

  • £136.51 / 100 tests


EU Stock 9
Add to basket

Description

Description

Description: The 5C3 monoclonal antibody reacts with human CD40, a 45-50kDa type I transmembrane glycoprotein. CD40 is a member of the TNFR family and is expressed by B lymphocytes, follicular dendritic cells, thymic epithelium, and a subset of peripheral T cells. CD40 regulates B cell development and maturation by inducing Ig isotype-switching and in combination with other signals such as IL-4, protects B cells from surface Ig-induced apoptosis and promotes proliferation. Interaction of CD40 with CD154 (gp39), its ligand on T cells, is important in T-B cell crosstalk and plays a role in costimulation and immune regulation. 5C3 is reported to be used for activation of B cells in in vitro functional assays.Applications Reported: The 5C3 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 5C3 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.015 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD40
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P25942
Bp50, CDW40, MGC9013, TNFRSF5, p50
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
5C3
PE
CD40
Conjugated
CD40
IgG1, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
958
Videos

For Research Use Only.