missing translation for 'onlineSavingsMsg'
Learn More

CD40, APC, clone: HM40-3, eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-0402-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15390600

  • £56.31 / 25µg


Estimated Shipment
30-07-2018
Add to basket

Description

Description

Description: The HM40-3 monoclonal antibody reacts with mouse and rat CD40, a 45-50kDa type I transmembrane glycoprotein. CD40 is a member of the TNFR family and is expressed by mouse B lymphocytes, follicular dendritic cells, thymic epithelium, and a subset of peripheral T cells. CD40 regulates B cell development/maturation by inducing Ig isotype switching and in combination with other signals such as IL-4, protects B cells from surface Ig-induced apoptosis and promotes proliferation. Interaction of CD40 with CD154 (gp39), its ligand on T cells, is important in T-B cell crosstalk and plays a role in costimulation and immune regulation.Applications Reported: This HM40-3 antibody has been reported for use in flow cytometric analysis.Applications Tested: This HM40-3 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD40
Monoclonal
0.2mg/mL
PBS with 0.09% sodium azide; pH 7.2
P27512
Bp50, CDW40, MGC9013, TNFRSF5, p50
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
MurineþRat
Flow Cytometry
HM40-3
APC
Cd40
Conjugated
Cd40
IgM, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
21939
Videos

For Research Use Only.