missing translation for 'onlineSavingsMsg'
Learn More

CD4, PE-Cyanine5, clone: GK1.5, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 15-0041-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15370160

  • £82.25 / 50µg


EU Stock 1
Add to basket

Description

Description

The GK1.5 monoclonal antibody reacts with the mouse CD4 molecule, a 55kDa cell surface receptor expressed by a majority of thymocytes, subpopulation of mature T cells and dendritic cells. CD4 binds to MHC class II on the surface of antigen presenting cells and plays an important role both in T cell development and in optimal functioning of mature T cells. In T cells, CD4 associates with protein tyrosine kinase p56lck through its cytoplasmic tail. Binding of GK1.5 is blocked by RM4-5.
Specifications

Specifications

CD4
GK1.5
PE-Cyanine5
Conjugated
Rat
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
L3T4, Ly-4
IgG2b, kappa
RUO
Primary
Murine
Videos

For Research Use Only.