missing translation for 'onlineSavingsMsg'
Learn More

CD4, FITC, clone: RM4-4, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-0043-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15247999

  • £49.65 / 50µg


Estimated Shipment date
from Supplier 29-01-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The RM4-4 monoclonal antibody reacts with the mouse CD4 molecule, a 55kDa cell surface receptor expressed by the majority of thymocytes, a subpopulation of mature T cells and dendritic cells. CD4 binds to MHC class II on the surface of antigen presenting cells and plays an important role both in T cell development and in optimal functioning of mature T cells. In T cells, CD4 associates with the protein tyrosine kinase lck through its cytoplasmic tail. Binding of RM4-4 does not block binding of the CD4 monoclonal antibodies RM4-5 or GK1.5.Applications Reported: This RM4-4 antibody has been reported for use in flow cytometric analysis.Applications Tested: This RM4-4 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD4
RM4-4
FITC
Conjugated
Rat
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
L3T4, Ly-4
IgG2b
RUO
Primary
Murine
Videos