missing translation for 'onlineSavingsMsg'
Learn More

CD4, eVolve 605, clone: RM4-5, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 83-0042-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15550737

  • £185.29 / 100 tests


EU Stock 11
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The RM4-5 monoclonal antibody reacts with the mouse CD4 molecule, a 55kDa cell surface receptor expressed by a majority of thymocytes, subpopulation of mature T cells and dendritic cells. CD4 binds to MHC class II on the surface of antigen presenting cells and plays an important role both in T cell development and in optimal functioning of mature T cells. In T cells, CD4 associates with protein tyrosine kinase p56lck through its cytoplasmic tail. Binding of RM4-5 is blocked by GK1.5.Applications Reported: This RM4-5 antibody has been reported for use in flow cytometric analysis.Applications Tested: This RM4-5 antibody has been pre-titrated and tested by flow cytometric analysis of mouse splenocytes. This can be used at 5 μL per test. A test is defined as the amount of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. For best results when using antibodies conjugated to eVolve™, we recommend a quick spin of the vial before use and avoid touching the bottom of the vial with the pipet tip.Laser/Filter Recommendation: eVolve™ 605 can be optimally excited with laser lines from 325-405 nm, and emits near 610 nm. We recommend using the violet (405 nm) laser line for excitation and a 595LP with a 605/40 or 610/20 bandpass filter, or equivalent, for detection.Fixation Recommendation: Antibodies conjugated to eVolve™ are compatible with eBioscience's fixation and permeabilization buffers.Excitation: 350-590 nm; Emission: 605 nm; Laser: Ultraviolet Laser, Violet LaserStorage and handling: For best results when using antibodies conjugated to eVolve™, we recommend a quick spin of the vial before use and avoid touching the bottom of the vial with the pipet tip.
Specifications

Specifications

CD4
RM4-5
eVolve 605
Conjugated
Rat
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material. This product should be used within 6 months from the date of receipt.
Monoclonal
Flow Cytometry
5μL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
L3T4, Ly-4
IgG2a
RUO
Primary
Murine
Videos