missing translation for 'onlineSavingsMsg'
Learn More

CD4, eFluor 450, clone: GK1.5, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-0041-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15321680

  • £140.71 / 100µg


EU Stock 19
Add to basket

Description

Description

Description: The GK1.5 monoclonal antibody reacts with the mouse CD4 molecule, a 55kDa cell surface receptor expressed by a majority of thymocytes, subpopulation of mature T cells and dendritic cells. CD4 binds to MHC class II on the surface of antigen presenting cells and plays an important role both in T cell development and in optimal functioning of mature T cells. In T cells, CD4 associates with protein tyrosine kinase p56lck through its cytoplasmic tail. Binding of GK1.5 is blocked by RM4-5.Applications Reported: This GK1.5 antibody has been reported for use in flow cytometric analysis.Applications Tested: This GK1.5 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD4
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P06332
CD4 antigen p55, CD4 receptor, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
GK1.5
eFluor 450
Cd4
Conjugated
Cd4
IgG2b, kappa
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
12504
Videos

For Research Use Only.