missing translation for 'onlineSavingsMsg'
Learn More

CD4, Biotin, clone: RM4-5, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0042-86

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15219419

  • £246.08 / 1mg


EU Stock 2
Add to basket

Description

Description

Description: The RM4-5 monoclonal antibody reacts with the mouse CD4 molecule, a 55kDa cell surface receptor expressed by a majority of thymocytes, subpopulation of mature T cells and dendritic cells. CD4 binds to MHC class II on the surface of antigen presenting cells and plays an important role both in T cell development and in optimal functioning of mature T cells. In T cells, CD4 associates with protein tyrosine kinase p56lck through its cytoplasmic tail. Binding of RM4-5 is blocked by GK1.5.Applications Reported: The RM4-5 antibody has been reported for use in flow cytometric analysis.Applications Tested: The RM4-5 antibody has been tested by flow cytometric analysis of mouse thymocytes and splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD4
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P06332
CD4 antigen p55, CD4 receptor, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
RM4-5
Biotin
Cd4
Conjugated
Cd4
IgG2a, kappa
1mg
Store at 2-8°C. Do not freeze.
Primary
12504
Videos

For Research Use Only.